Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM29A Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen FAM29A
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093684
View Documents
Novus Biologicals
NBP303788100UL
100 μg Each for $320.71
Add to cart
 
NB093685
View Documents
Novus Biologicals
NBP30378820UL
20 μg Each for $138.37
Add to cart
 
Description

Description

FAM29A Polyclonal antibody specifically detects FAM29A in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

FAM29A
Unconjugated
Rabbit
Human, Mouse
Dgt6, dim gamma-tubulin homolog, FAM29AMGC102696, family with sequence similarity 29, member A, FLJ20060, HAUS augmin-like complex subunit 6, HAUS augmin-like complex, subunit 6, KIAA1574, MGC138798, MGC138799
Recombinant fusion protein containing a sequence corresponding to amino acids 676-955 of human FAM29A (NP_060115.3).
Polyclonal
Affinity purified
Immunofluorescence, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
54801
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit