Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIEZO2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548758

Catalog No. PA548758

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Component of mechanosensitive channel required for rapidly adapting mechanically activated (MA) currents. Requires for Merkel-cell mechanotransduction.
Specifications

Specifications

PIEZO2
Polyclonal
PBS with 0.09% sodium azide
Q9H5I5
Piezo-type mechanosensitive ion channel component 2, Protein FAM38B, PIEZO2, C18orf30, C18orf58, FAM38B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
63895
Western Blot
Unconjugated
PIEZO2
Liquid
PIEZO2
KLH conjugated synthetic peptide between 2575-2604 amino acids from the N-terminal region of human FAM38B.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit