Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM44B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557600
Description
Immunogen sequence: PINPASLPPG DPQLIALIVE QLKSR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 92%.
FAM44B required for proper chromosome biorientation through the detection or correction of syntelic attachments in mitotic spindles.
Specifications
FAM44B | |
Polyclonal | |
Unconjugated | |
BOD1 | |
2310022M17Rik; biorientation defective 1; biorientation defective protein 1; biorientation of chromosomes in cell division 1; biorientation of chromosomes in cell division protein 1; BOD1; BOD2.1; bone length and organs 1; Fam44b; family with sequence similarity 44, member B; Protein FAM44B; RGD1305622 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91272 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96IK1 | |
BOD1 | |
Recombinant protein corresponding to Human FAM44B. Recombinant protein control fragment (Product #RP-96893). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction