Learn More
FAM50A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 198491AP
Catalog No. 50-173-2573
Add to cart
Description
Description
May be a DNA-binding protein or transcriptional factor.
Specifications
Specifications
FAM50A
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14320, Q9WV03
9F, DXS9928E, FAM50A, HXC 26, HXC26, Protein FAM50A, Protein HXC 26, Protein XAP 5, XAP5
Rabbit
IgG
150 μL
-20°C
Primary
108160, 293862, 9130
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
FAM50A
Liquid
FAM50A
FAM50A Fusion Protein Ag13907
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat