Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM57B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
$496.07
Specifications
Antigen | FAM57B |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3067
|
Proteintech Group Inc
207601AP150UL |
150 μL |
Each of 1 for $496.07
|
|
|||||
Description
FAM57B is a protein coding gene.Specifications
FAM57B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q71RH2, Q7TNV1 | |
293493, 68952, 83723 | |
FAM57B Fusion Protein Ag14707 | |
Primary | |
-20°C | |
FAM57B |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FAM57B, FP1188, Protein FAM57B | |
Fam57b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title