Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM60A Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen FAM60A
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125243
View Documents
Novus Biologicals
NBP310171100UL
100 μg
Each for $352.89
Add to cart
 
NB125244
View Documents
Novus Biologicals
NBP31017125UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

FAM60A Polyclonal antibody specifically detects FAM60A in Mouse samples. It is validated for Western Blot
Specifications

Specifications

FAM60A
Western Blot
Purified
RUO
Mouse
C12orf14, chromosome 12 open reading frame 14, family with sequence similarity 60, member A, TERA, Tera protein homolog
The immunogen is a synthetic peptide directed towards the N terminal region of mouse FAM60A (NP_062617.2). Peptide sequence SSSRFTDSKRYEKDFQSCFGLHETRSGDICNACVLLVKRWKKLPAGSKKN
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS buffer, 2% sucrose
58516
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit