Learn More
FAM62A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 214501AP
Catalog No. 50-173-3426
Add to cart
Description
Description
ESYT1 gene ontology annotations related to this gene include endoplasmic reticulum-plasma membrane tethering; lipid transport.
Specifications
Specifications
FAM62A
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3U7R1, Q9BSJ8, Q9Z1X1
E Syt1, ESYT1, Extended synaptotagmin 1, FAM62A, KIAA0747, MBC2
Rabbit
IgG
150 μL
-20°C
Primary
23344, 23943, 29579
Immunoprecipitation, Western Blot
Unconjugated
ESYT1
Liquid
ESYT1
FAM62A Fusion Protein Ag13895
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat