Learn More
Aviva Systems Biology Corp FAM62C Antibody - middle region (ARP50020_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50020_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM62C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM62C. Alternate names: CHR3SYT, E-Syt3, FAM62CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60242