Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM65A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552228
Description
Immunogen sequence: HALESFSFLN EDEDEDNDVP GDRPPSSPEA GAEDSIDSPS ARPLSTGCPA LDAALVRHLY HCSRLLLKLG TFGPLRCQEA WALERLLREA RVLEAVCEFS RRWEIPASSA QEVVQFS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
Downstream effector protein for Rho-type small GTPases that plays a role in cell polarity and directional migration (PubMed:27807006). Acts as an adapter protein, linking active Rho proteins to STK24 and STK26 kinases, and hence positively regulates Golgi reorientation in polarized cell migration upon Rho activation (PubMed:27807006). Involved in the subcellular relocation of STK26 from the Golgi to cytoplasm punctae in a Rho- and PDCD10-dependent manner upon serum stimulation (PubMed:27807006). [UniProt].
Specifications
FAM65A | |
Polyclonal | |
Unconjugated | |
RIPOR1 | |
2310066E14Rik; AI837433; Fam65a; family with sequence similarity 65 member A; family with sequence similarity 65, member A; FLJ13725; Kiaa1930; protein FAM65A; Rho family-interacting cell polarization regulator 1; RIPOR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZS17 | |
RIPOR1 | |
Recombinant protein corresponding to Human FAM65A. Recombinant protein control fragment (Product #RP-100912). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction