Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM65A Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30437120UL
Description
FAM65A Polyclonal specifically detects FAM65A in Mouse samples. It is validated for Western Blot.Specifications
FAM65A | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
79567 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
family with sequence similarity 65, member A, hypothetical protein LOC79567 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1004-1223 of human FAM65A (NP_001180451.1). | |
20 μL | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction