Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM65B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548820

Catalog No. PA548820


Add to cart

Description

Description

Required for hearing (PubMed:24958875). Involved in skeletal muscle development (PubMed:24687993).
Specifications

Specifications

FAM65B
Polyclonal
PBS with 0.09% sodium azide
Q9Y4F9
Protein FAM65B, FAM65B, C6orf32, DIFF48, KIAA0386, PL48
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9750
Western Blot
Unconjugated
RIPOR2
Liquid
RIPOR2
KLH conjugated synthetic peptide between 829-856 amino acids from the C-terminal region of human FAM65B.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit