Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM69B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548157

Catalog No. PA548157

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the FAM69 family of cysteine-rich type II transmembrane proteins. These proteins localize to the endoplasmic reticulum but their specific functions are unknown. [provided by RefSeq, Nov 2011]
Specifications

Specifications

FAM69B
Polyclonal
PBS with 0.09% sodium azide
Q5VUD6, Q99ML4
Protein FAM69B, FAM69B, C9orf136
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
138311, 56279
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FAM69B
Liquid
FAM69B
KLH conjugated synthetic peptide between 107-136 amino acids from the N-terminal region of human FAM69B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit