Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM73A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532001
Description
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
FAM73A | |
Polyclonal | |
Unconjugated | |
MIGA1 | |
FAM73A; family with sequence similarity 73 member A; family with sequence similarity 73, member A; MIGA1; Mitoguardin 1; mitoguardin-1; protein FAM73A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
374986 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NAN2 | |
MIGA1 | |
Recombinant fragment corresponding to a region within amino acids 401 and 632 of Human FAM73A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction