Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM76A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155436
Description
FAM76A Polyclonal specifically detects FAM76A in Human samples. It is validated for Western Blot.Specifications
FAM76A | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q8TAV0 | |
FAM76A | |
Synthetic peptides corresponding to FAM76A(family with sequence similarity 76, member A) The peptide sequence was selected from the middle region of FAM76A. Peptide sequence QCAFDRKDDRKKVDGKLLCWLCTLSYKRVLQKTKEQRKHLSSSSRAGHQE. | |
Affinity Purified | |
RUO | |
199870 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
family with sequence similarity 76, member A, FLJ41946, hypothetical protein LOC199870, MGC34648 | |
Rabbit | |
35 kDa | |
100 μL | |
Primary | |
Zebrafish: 80%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title