Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM78B Polyclonal Antibody
FAM78B Polyclonal Antibody
Supplier: Thermo Scientific PA562684
Description
FAM78B Polyclonal Antibody for IHC

Specifications
FAM78B | |
Polyclonal | |
Unconjugated | |
FAM78B | |
C030014K22Rik; C030020L09Rik; Fam78b; family with sequence similarity 78 member B; family with sequence similarity 78, member B; protein FAM78B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
149297 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VT40 | |
FAM78B | |
Recombinant protein corresponding to Human FAM78B. Recombinant protein control fragment (Product #RP-101701). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction