Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM82A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557024
Description
Immunogen sequence: SFPVPKAFNT RVEELNLDVL LQKVDHLRMS ESGKSESFEL LRDHKEKFRD EIEFMWRFAR AYGDMYELST NTQEKK Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 78%.
The human protein FAM82A1, also known as regulator of microtubule dynamics 2 (RMD2), was one of three proteins identified as related to a family of microtubule-associated proteins in C. elegans. FAM82A1 contains multiple coiled-coil domains and localize to the spindle microtubules and spindle poles during cell division. During interphase, RMD2 localizes to the cytoplasm with protein observed in the microtubule lattice and perinuclear region.
Specifications
FAM82A1 | |
Polyclonal | |
Unconjugated | |
RMDN2 | |
AW061290; BLOCK18; Fam82a; Fam82a1; family with sequence similarity 82, member A; family with sequence similarity 82, member A1; microtubule-associated protein; mRMD-2; PRO34163; Protein FAM82A1; PYST9371; regulator of microtubule dynamics 2; regulator of microtubule dynamics protein 2; RMD2; RMD-2; RMD4; Rmdn2; UNQ9371/PRO34163 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151393 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LZ7 | |
RMDN2 | |
Recombinant protein corresponding to Human FAM82A1. Recombinant protein control fragment (Product #RP-102630). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction