Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM83E Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19148520UL

 View more versions of this product

Catalog No. NBP19148520


Add to cart

Description

Description

FAM83E Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

FAM83E
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
FAM83E
Synthetic peptide directed towards the middle region of human FAM83E. Peptide sequence RARTPSGPPARPSRSMWDLSRLSQLSGSSDGDNELKKSWGSKDTPAKALM.
Immunogen affinity purified
RUO
Primary
54854
Western Blot
Western Blot 1:1000
NP_060178
family with sequence similarity 83, member E, FLJ20200, hypothetical protein LOC54854, MGC138175, MGC138177
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit