Learn More
Invitrogen™ FAM83F Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113969
Description
Immunogen sequence: KDEKAPHLKQ VVRQMIQQAQ KVIAVVMDLF TDGDIFQDIV DAACKRRVPV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
FAM83F is a protein coding gene.
Specifications
FAM83F | |
Polyclonal | |
Unconjugated | |
FAM83F | |
AW544981; FAM83F; family with sequence similarity 83 member F; family with sequence similarity 83, member F; protein FAM83F; RGD1562089 | |
Rabbit | |
Affinity chromatography | |
RUO | |
113828 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NEG4 | |
FAM83F | |
Synthetic peptide directed towards the N terminal region of human FAM83F (aa 143-192). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.