Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM83F Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | FAM83F |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179503
|
Novus Biologicals
NBP179503 |
100 μL |
Each of 1 for $436.00
|
|
Description
FAM83F Polyclonal specifically detects FAM83F in Human samples. It is validated for Western Blot.Specifications
FAM83F | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
family with sequence similarity 83, member F, hypothetical protein LOC113828 | |
FAM83F | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_612444 | |
113828 | |
Synthetic peptide directed towards the N terminal of human FAM83FThe immunogen for this antibody is FAM83F. Peptide sequence KDEKAPHLKQVVRQMIQQAQKVIAVVMDLFTDGDIFQDIVDAACKRRVPV. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title