Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM83G Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309714100UL
Description
FAM83G Polyclonal specifically detects FAM83G in Human samples. It is validated for Western Blot.Specifications
FAM83G | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
family with sequence similarity 83, member G, FLJ41564 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human FAM83G (NP_001035088). Peptide sequence DGRATEEHPSPFGIPYSKLSQSKHLKARTGGSQWASSDSKRRAQAPRDRK | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
644815 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction