Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM86A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546445
Description
Peptide sequence: CPEAIMSLVG VLRRLAACRE HQRAPEVYVA FTVRNPETCQ LFTTELGRAG Sequence homology: Human: 100%.
Catalyzes the trimethylation of eukaryotic elongation factor 2 (EEF2) on 'Lys-525'. [UniProt].
Specifications
FAM86A | |
Polyclonal | |
Unconjugated | |
EEF2KMT | |
EEF2KMT; eEF2-KMT; eEF2-lysine methyltransferase; eukaryotic elongation factor 2 lysine methyltransferase; FAM86A; family with sequence similarity 86, member A; protein FAM86A; protein-lysine N-methyltransferase EEF2KMT; putative protein N-methyltransferase FAM86A; SB153 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
196483 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96G04 | |
EEF2KMT | |
Synthetic peptide directed towards the C-terminal of human FAM86A (aa 252-301). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction