Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM90A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544694
Description
Peptide sequence: PDEEDPRLKC KNCEAFGHTA RSTRCPMKCW KAALVPPNFG EKEGKENLKP Sequence homology: Human: 100%.
FAM90A1 belongs to subfamily I of the primate-specific FAM90A gene family, which originated from multiple duplications and rearrangements (Bosch et al., 2007 [PubMed 17684299]).
Specifications
FAM90A1 | |
Polyclonal | |
Unconjugated | |
FAM90A1 | |
FAM90A1; family with sequence similarity 90 member A1; protein FAM90A1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55138 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86YD7 | |
FAM90A1 | |
Synthetic peptide directed towards the N-terminal of human FAM90A1 (aa 35-84). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction