Learn More
Aviva Systems Biology Corp FAM98A Antibody - middle region (ARP55265_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55265_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM98A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM98A. Alternate names: DKFZP564F0522, DKFZp686O03192ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61450