Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM98A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557584
Description
Immunogen sequence: LLTYLISELE AARMLCVNAP PKKAQEGGGS EVFQELKGIC IALGMSKPPA NITMFQFFSG IEKKLKETLA KVPPNH Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
FAM98A belongs to the FAM98 family. The function of FAM98A remains unknown.
Specifications
FAM98A | |
Polyclonal | |
Unconjugated | |
Fam98a | |
2810405J04Rik; AL024114; Fam98a; FAM98A-like protein; family with sequence similarity 98 member A; family with sequence similarity 98, member A; protein FAM98A; RGD1305486 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25940 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCA5 | |
Fam98a | |
Recombinant protein corresponding to Human FAM98A. Recombinant protein control fragment (Product #RP-96054). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction