Learn More
Abnova™ FAM9B 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00171483T01
Catalog No. 89-944-034
Add to cart
Specifications
Specifications
Western Blot
293T
NM_205849
pCMV-FAM9B full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
FAM9B 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 22.4kDa
Transient overexpression cell lysate was tested with anti-FAM9B antibody (Mfr. No. H00171483-B03) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)