Learn More
Lab-Aids™ Families of Elements Experiment Kit
Learn to classify elements by observing behavior of their compounds, examine properties of the halogen compounds and determine formulas of compounds through experimentation.
Supplier: Lab-Aids™ 110R
Description
Students learn how to classify elements by observing the behavior of compounds in the Halogen family (Group VIIA). The chemical properties of this group are very similar despite the marked differences in their physical properties. As students perform a series of experiments, they discover that the elements form remarkably similar compounds with silver. Also, that the formulas of the compounds are similar, and that the elements are indeed in the same family.
Refill includes:
3 Drop control bottles - Potassium Chloride
3 Drop control bottles - Potassium Bromide
3 Drop control bottles - Potassium Iodide
3 Drop control bottles - Potassium Sulfide
9 Drop control bottles - Silver Nitrate
Order Info
Class size: 30 students.
Specifications
To explore the chemical properties of the halogen family (group viia) | |
15 chemplates™ with spatula, drop controlled bottles of 3 potassium chloride, 3 potassium bromide, 3 potassium iodide and 3 potassium sulfide, 9 silver nitrate, 60 student worksheets and guides and instruction manual with msds | |
Families Of Elements Experiment Kit | |
30 Students |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.