Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Famotidine, USP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
76824-35-6
C8H15N7O2S3
337.44
XUFQPHANEAPEMJ-UHFFFAOYSA-N
3-[({2-[(diaminomethylidene)amino]-1,3-thiazol-4-yl}methyl)sulfanyl]-N'-sulfamoylpropanimidamide
NC(N)=NC1=NC(CSCCC(N)=NS(N)(=O)=O)=CS1
76824-35-6
C8H15N7O2S3
337.44
XUFQPHANEAPEMJ-UHFFFAOYSA-N
3-[({2-[(diaminomethylidene)amino]-1,3-thiazol-4-yl}methyl)sulfanyl]-N'-sulfamoylpropanimidamide
NC(N)=NC1=NC(CSCCC(N)=NS(N)(=O)=O)=CS1

F1154, 76824-35-6, C8H15N7O2S3

Spectrum™ Famotidine, USP is a histamine and H2 receptor antagonist used to treat peptic ulcers and heartburn. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 1 g
CAS Max % 1
Percent Purity 98.5 to 101%