Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Famotidine, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog Number 18602600
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
76824-35-6
C8H15N7O2S3
337.44
XUFQPHANEAPEMJ-UHFFFAOYSA-N
3-[({2-[(diaminomethylidene)amino]-1,3-thiazol-4-yl}methyl)sulfanyl]-N'-sulfamoylpropanimidamide
NC(N)=NC1=NC(CSCCC(N)=NS(N)(=O)=O)=CS1
76824-35-6
C8H15N7O2S3
337.44
XUFQPHANEAPEMJ-UHFFFAOYSA-N
3-[({2-[(diaminomethylidene)amino]-1,3-thiazol-4-yl}methyl)sulfanyl]-N'-sulfamoylpropanimidamide
NC(N)=NC1=NC(CSCCC(N)=NS(N)(=O)=O)=CS1

F1154, 76824-35-6, C8H15N7O2S3

Spectrum™ Chemical Famotidine, USP is a histamine and H2 receptor antagonist used to treat peptic ulcers and heartburn. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98.5 to 101%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit