Learn More
Fisherbrand™ Fan Blower Switch for 2 HP fan
Fisherbrand™ Fittings for Fume Hoods are designed to be used with any Fisherbrand Fume Hood
Catalog No. 06-000-605
Add to cart
Description
Description
  • Manual fan motor starter switch, toggle style, single pole, with red pilot light, stainless steel cover plate and 1 thermal unit.
  • Fixtures shipped loose for installation at the job site by mechanical trades.
Specifications
Specifications
2 HP fan