Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fananserin, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
127625-29-0
C23H24FN3O2S
425.522
VGIGHGMPMUCLIQ-UHFFFAOYSA-N
60785
C1CN(CCN1CCCN2C3=CC=CC4=C3C(=CC=C4)S2(=O)=O)C5=CC=C(C=C5)F
This item is not returnable. View return policy
127625-29-0
C23H24FN3O2S
425.522
VGIGHGMPMUCLIQ-UHFFFAOYSA-N
60785
C1CN(CCN1CCCN2C3=CC=CC4=C3C(=CC=C4)S2(=O)=O)C5=CC=C(C=C5)F

5-HT2A antagonist. Also D4 antagonist

5-HT2A receptor antagonist that also displays high affinity at D4 receptors (Ki values are 0.26, 2.93, 25, 38, 70 and 726 nM for 5-HT2A, D4, H1, α1, 5-HT1A and D2 receptors respectively). Inhibits inositol phosphate formation evoked by 5-HT in vitro (IC50 = 7.76 nM) and antagonizes mescaline-induced head twitches in vivo.
Quantity 10 mg
Synonym fananserin, fananserin usan:inn, 3h fananserin, unii-38qj762et6, fananserin usan/inn, chembl83894, 2-3-4-4-fluorophenyl-1-piperazinyl propyl-2h-naphth 1,8-cd isothiazole 1,1-dioxide, 2-3-4-p-fluorophenyl-1-piperazinyl propyl-2h-naphth 1,8-cd isothiazole 1,1-dioxide, 2h-naphth 1,8-cd isothiazole, 2-3-4-4-fluorophenyl-1-piperazinyl propyl-, 1,1-dioxide, dsstox_cid_26743
Formula Weight 425.52
Percent Purity >99%
Chemical Name or Material Fananserin

Recommended Storage : Store at Room Temperature