Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCC Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA123348

Catalog No. PIPA123348


Add to Cart

Description

Description

Recommended positive controls: HeLa nuclear cell lysate.

FANCC Polyclonal antibody specifically detects FANCC in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FANCC
Polyclonal
Unconjugated
FANCC
FA3; Fac; Facc; FANCC; Fanconi anemia complementation group C; Fanconi anemia group C protein; Fanconi anemia group C protein homolog; Fanconi anemia, complementation group C; FLJ14675; mir-3074-1; Protein FACC; RP11-80I15.2
Rabbit
Antigen affinity chromatography
RUO
2176
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with no preservative; pH 7.2
Q00597
FANCC
A synthetic peptide corresponding to the C-terminal 15 amino acids, conjugated to a carrier protein
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit