Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCD2 Antibody (1290C), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP261930APC
Description
FANCD2 Monoclonal antibody specifically detects FANCD2 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDownSpecifications
FANCD2 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, KnockDown | |
1290C | |
Western Blot, Knockout Validated | |
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
This FANCD2 Antibody (1290C) was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] | |
0.1 mL | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
2177 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction