Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCD2 Rabbit anti-Human, Biotin, Clone: 1290C, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261930B
Description
FANCD2 Monoclonal antibody specifically detects FANCD2 in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
FANCD2 | |
1290C | |
Western Blot, Knockout Validated | |
PBS with 0.05% Sodium Azide | |
2177 | |
Rabbit | |
IgG | |
Primary | |
0.1mL | |
Store at 4°C in the dark. |
KnockDown,Western Blot | |
Biotin | |
Purified | |
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
FANCD2 | |
This FANCD2 Antibody (1290C) [Biotin] was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] | |
Monoclonal | |
Protein A or G purified | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
Human,Mouse,Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title