Learn More
FANCD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 240061AP
Catalog No. 50-173-4203
Add to cart
Description
Description
The antibody recognize both un-phospho and phospho-Ser(330) of FANCD2.
Fanconi anemia (FANC) is a human autosomal-recessive cancer susceptibility disorder characterized by congenital defects, progressive bone marrow failure, and cellular hypersensitivity to mitomycin C (MMC). The FANC subunit D2 protein is vital for cellular resistance to DNA cross-linking and the arrest of DNA synthesis after ionizing radiation. DNA damage activates the monoubiquitination of FANCD2, targeting nuclear foci containing the BRCA1 protein.
Specifications
Specifications
FANCD2
0.44 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80V62, Q9BXW9
FA D2, FA4, FACD, FAD, FAD2, FANCD, FANCD2, Protein FACD2
Rabbit
IgG
150 μL
-20°C
Primary
211651, 2177
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
FANCD2
Liquid
Fancd2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse