Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2 Rabbit anti-Human, PE/Cy5.5, Clone: 1290D, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254808PECY55

Catalog No. N254808PEC5


Add to cart

Description

Description

FANCD2 Monoclonal antibody specifically detects FANCD2 in Human samples. It is validated for Flow Cytometry, Immunoassay.
Specifications

Specifications

FANCD2
1290D
Flow Cytometry, Knockout Validated
Conjugated, Purified, Supernatant
FANCD2
Human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot No.Q9BXW9]
Protein A or G purified from cell culture supernatant
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Phospho Specific
Primary
2177.0
Flow Cytometry, Immunoassay
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
DKFZp762A223, FA4, FACDFLJ23826, FA-D2, FADFAD2, FANCD, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, Protein FACD2
Rabbit
IgG
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit