Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fancd2os Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569927
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
FANCD2OS is a protein coding gene.
Specifications
Fancd2os | |
Polyclonal | |
Unconjugated | |
FANCD2OS | |
4931417G12Rik; C20H3orf24; C22H3orf24; C3orf24; Fancd2 opposite strand; FANCD2 opposite strand protein; FANCD2OS; Fanconi anemia group D2 protein opposite strand transcript protein; HSD19; RGD1565997; uncharacterized protein C3orf24 homolog; Unknown (protein for MGC:133872) | |
Rabbit | |
Affinity chromatography | |
RUO | |
115795 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96PS1 | |
FANCD2OS | |
synthetic peptide directed towards the middle region of human C3orf24. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction