Learn More
Abnova™ FANCL 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00055120T02
Catalog No. 89-944-059
Add to cart
Specifications
Specifications
Western Blot
293T
NM_018062.2
pCMV-FANCL full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
FANCL 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 41.36kDa
Transient overexpression cell lysate was tested with anti-FANCL antibody (Mfr. No. H00055120-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)