Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$337.75 - $627.50
Specifications
Antigen | FANCL |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FANCL Polyclonal specifically detects FANCL in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FANCL | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
E3 ubiquitin-protein ligase FANCL, EC 6.3.2.-, FAAP43PHF9FLJ10335, Fanconi anemia group L protein, Fanconi anemia, complementation group L, Fanconi anemia-associated polypeptide of 43 kDa, PHD finger protein 9, Pog | |
FANCL | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
55120 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FPARAILEKSDFTMDCGICYAYQLDGTIPDQVCDNSQCGQPFHQICLYEWLRGLLTSRQSFNIIFGECPYCSKPIT | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title