Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FARSB Polyclonal Antibody
FARSB Polyclonal Antibody
Supplier: Thermo Scientific PA530350
Description
FARSB Polyclonal Antibody for Western Blot, IHC (P)

Specifications
FARSB | |
Polyclonal | |
Unconjugated | |
Farsb | |
C76708; Farsa; FARSB; Farsl; Farslb; Frsb; HSPC173; ph; PheHB; phenylalanine tRNA ligase 1, beta, cytoplasmic; phenylalanine tRNA synthetase alpha; phenylalanine-tRNA ligase beta chain; phenylalanine--tRNA ligase beta chain; phenylalanine--tRNA ligase beta subunit; phenylalanine-tRNA synthetase-like, beta subunit; phenylalanyl-tRNA synthetase beta chain; phenylalanyl-tRNA synthetase beta subunit; phenylalanyl-tRNA synthetase beta-subunit; phenylalanyl-tRNA synthetase, beta subunit; phenylalanyl-tRNA synthetase-like, beta subunit; PheRS; PheRS alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10056, 23874, 301544 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NSD9, Q9WUA2 | |
Farsb | |
Recombinant fragment corresponding to a region within amino acids 378 and 589 of Human FARSB. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction