Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Fas Ligand, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Fas Ligand, Rabbit Polyclonal Antibody.

Supplier:  Epredia™ RB9029R1

Catalog No. RB9029R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Fas ligand is a cell surface molecule belonging to the tumor necrosis factor family,. It binds to its receptor Fas, thus inducing apoptosis. Various cells express Fas, where Fas ligand is expressed predominantly on activated T cells. Fas and Fas ligand are involved in the down-regulation of immune reactions as well as T cell-mediated cytotoxicity.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminal of human Fas ligand

Molecular Weight: 37 kDa

Positive Control: Tonsil

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Fas Ligand
Polyclonal
Rabbit
1 mL
Apoptosis
Human, Mouse
IgG
Western Blot
Unconjugated
A synthetic peptide derived from N-terminal of human Fas ligand
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit