Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAS Mouse anti-Human, Unconjugated, Clone: B-R18, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB21097
Description
FAS Monoclonal antibody specifically detects FAS in Human samples. It is validated for IHC-P, IF, Flow Cyt.Specifications
FAS | |
B-R18 | |
Fas (TNF receptor superfamily, member 6) | |
Liquid | |
Mouse | |
IgG1, Kappa | |
Store at 2 to 8°C | |
Monoclonal | |
Human |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
FAS | |
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6 | |
Recombinant protein corresponding to human FAS | |
100 μg | |
Primary | |
355 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title