Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAS Mouse anti-Human, Unconjugated, Clone: B-R18, Abnova™

Mouse Monoclonal Antibody.

Manufacturer:  Abnova Corporation MAB21098

Catalog No. 89-337-591


Add to cart

Description

Description

FAS Monoclonal antibody specifically detects FAS in Human samples. It is validated for IHC-P, IF, Flow Cyt.
Specifications

Specifications

FAS
B-R18
Fas (TNF receptor superfamily, member 6)
Liquid
Mouse
IgG1, Kappa
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
FAS
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
Recombinant protein corresponding to human FAS
100 μg
Primary
355
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit