Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas/TNFRSF6/CD95, Human anti-Human, Clone: R-125224, Novus Biologicals™

Human Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen Fas/TNFRSF6/CD95
Clone R-125224
Applications Flow Cytometry, ELISA, Functional Assay
Host Species Human
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016346
View Documents
Novus Biologicals
NBP2811120.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB016347
View Documents
Novus Biologicals
NBP2811120.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Fas/TNFRSF6/CD95 Monoclonal antibody specifically detects Fas/TNFRSF6/CD95 in Human Samples. It is validated for Flow Cytometry, ELISA, Functional.
Specifications

Specifications

Fas/TNFRSF6/CD95
Flow Cytometry, ELISA, Functional Assay
Human
Primary
R-125224
Human
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit