Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas/TNFRSF6/CD95 Mouse anti-Human, Clone: 50825, R&D Systems™

Mouse Monoclonal Antibody

$116.82 - $320.50

Specifications

Antigen Fas/TNFRSF6/CD95
Clone 50825
Dilution Flow Cytometry 0.25 ug/10^6 cells
Applications Flow Cytometry
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB326100 View Documents R&D Systems
MAB326100
100 ug Each for $320.50
Add to cart
 
MAB326SP View Documents R&D Systems
MAB326SP
25 ug Each for $116.82
Add to cart
 
Description

Description

Fas/TNFRSF6/CD95 Monoclonal antibody specifically detects Fas/TNFRSF6/CD95 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Fas/TNFRSF6/CD95
Flow Cytometry 0.25 ug/10^6 cells
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Apo-1, Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1, APT1FASTM, CD95, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Mouse myeloma cell line NS0-derived recombinant human Fas/TNFRSF6/CD95 Gln26-Asn173 Accession # P25445
Monoclonal
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
50825
Flow Cytometry
Purified
Human
P25445
355
IgG1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Fas/TNFRSF6/CD95 in direct ELISAs.

RUO

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit