Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fascin-3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549583

Catalog No. PA549583


Add to cart

Description

Description

Acts as an actin bundling protein.
Specifications

Specifications

Fascin-3
Polyclonal
PBS with 0.09% sodium azide
Q9NQT6, Q9QXW4
Fascin-3, Testis fascin, FSCN3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29999, 56223
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FSCN3
Liquid
FSCN3
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human Fascin-3.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit