Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Fascin Recombinant Rabbit Monoclonal Antibody (1B3V6)
Fascin Recombinant Rabbit Monoclonal Antibody (ARC1638)
Supplier: Invitrogen™ MA535811
Description
Fascin Recombinant Monoclonal Antibody for Western Blot
Fascin organizes filamentous actin into bundles with a minimum of 4. 1:1 actin/fascin ratio. It is probably involved in the assembly of actin filament bundles present in microspikes, membrane ruffles, and stress fibers.
Specifications
Fascin | |
Recombinant Monoclonal | |
0.28 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P85845, Q16658, Q61553 | |
FSCN1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 249-362 of human Fascin/FSCN1 (Q16658). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1B3V6 | |
Unconjugated | |
FSCN1 | |
55 kDa actin-bundling protein; 55kDa actin-bundling protein; AI663989; Fan1; fascin; fascin 1; fascin actin-bundling protein 1; Fascin homolog 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus); fascin homolog 1, actin-bundling protein; Fascin1; fascin-1; Fscn1; fscn1 {ECO:0000250; HSN; p55; Singed; Singed (Drosophila) like (sea urchin fascin homolog like); singed-like (fascin homolog, sea urchin); singed-like protein; SNL; UniProtKB:Q16658} | |
Rabbit | |
Affinity chromatography | |
RUO | |
14086, 6624, 683788 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction