Learn More
ACROBiosystems Recombinant Protein;Human Fas / TNFRSF6 / CD95 Protein;HEK293;50UG;FAS,FAS,ALPS1A,APO-1,APO1,APT1,APT-1,CD95,CD-95,FAS1,FAS-1,FASTM,TNFRSF6,TNFRSF-6,FasR ,Fas-R
Supplier:  ACROBiosystems FASH522950UG
Encompass_Preferred
Recombinant Protein;Human Fas / TNFRSF6 / CD95 Protein;50UG;Human Fas, His Tag (FAS-H5229) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Asn 173 (Accession # AAH12479.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.2 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7578
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.