Learn More
Abnova™ FASLG (Rat) ELISA Kit
Sandwich enzyme-linked immunosorbent assay
Manufacturer:  Abnova™ KA5720
Catalog No. 89-339-696
Add to cart
Description
Description
FASLG (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat FASL in cell culture supernates, serum and plasma (heparin, EDTA, citrate).
Specifications
Specifications
25385
Standard curve
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
Apt1Lg1, Fasl, Tnfsf6
Faslg
1 Kit
RUO
Rat