Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Violet B salt, 90%, purified, ACROS Organics™

Click to view available options
Quantity:
25g
14726-28-4
C15H14ClN3O2·1/2ZnCl2
MFCD00083330
4-Benzamido-2-methoxy-5-methylbenzenediazonium trichlorozincate,Azoene fast Violet B salt,C.I. 37165,Azoic Diazo No. 41
14726-28-4
C15H14ClN3O2·1/2ZnCl2
MFCD00083330
4-Benzamido-2-methoxy-5-methylbenzenediazonium trichlorozincate,Azoene fast Violet B salt,C.I. 37165,Azoic Diazo No. 41
Assay Percent Range 90%
Infrared Spectrum Authentic
Loss on Drying 1% max.
Formula Weight 371.88
Physical Form Crystalline Powder
Percent Purity ≥88% (ex N)
Color Beige-Yellow to Brown
Quantity 25g
Chemical Name or Material Fast violet B salt, 90%

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 238-775-4