Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Yellow GC Salt, Acros Organics

Click to view available options
Quantity:
25g
14263-92-4
C12H8Cl6N4Zn
486.304
DAGOHHHQXOCGSB-UHFFFAOYSA-J
Azoic Diazo No. 44,C.I. 37000
167040
zinc;2-chlorobenzenediazonium;tetrachloride
C1=CC=C(C(=C1)[N+]#N)Cl.C1=CC=C(C(=C1)[N+]#N)Cl.[Cl-].[Cl-].[Cl-].[Cl-].[Zn+2]
14263-92-4
C12H8Cl6N4Zn
486.304
DAGOHHHQXOCGSB-UHFFFAOYSA-J
Azoic Diazo No. 44,C.I. 37000
167040
zinc;2-chlorobenzenediazonium;tetrachloride
C1=CC=C(C(=C1)[N+]#N)Cl.C1=CC=C(C(=C1)[N+]#N)Cl.[Cl-].[Cl-].[Cl-].[Cl-].[Zn+2]
Formula Weight 311.29
Quantity 25g
Chemical Name or Material Fast Yellow GC salt